Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282801.1 | NP_001269730.1 | 4, 5 | 403 | ||
NM_001282802.1 | NP_001269731.1 | 4, 5 | 551 | |||
NM_020654.4 | NP_065705.3 | 5, 6 | 601 | |||
XM_011513038.2 | XP_011511340.1 | 5, 6 | 590 | |||
XM_011513039.2 | XP_011511341.1 | 4, 5 | 515 | |||
XM_011513041.2 | XP_011511343.1 | 4, 5 | 392 | |||
XM_011513045.2 | XP_011511347.1 | 5, 6 | 591 | |||
XM_017006924.1 | XP_016862413.1 | 5, 6 | 478 | |||
XM_017006925.1 | XP_016862414.1 | 5, 6 | 566 | |||
XM_017006927.1 | XP_016862416.1 | 3, 4 | 301 | |||
XM_017006928.1 | XP_016862417.1 | 5, 6 | 591 | |||
GenBank mRNA | AF199458.1 | - | 4, 5 | 549 | ||
AK297356.1 | - | 512 | ||||
AL136599.1 | - | 4, 5 | 429 | |||
BC062674.1 | - | 5, 6 | 548 | |||
BC129988.1 | - | 5, 6 | 528 | 40128878 |