Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other USP28 SIRNA Assays ›
Gene Symbol:
USP28
Gene Name:
ubiquitin specific peptidase 28
Gene Aliases:
-
Chromosome Location:
Chr.11: 113797875 - 113875580 on Build GRCh38
UniGene ID:
Hs.503891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
105210
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: USP28
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301029.1
NP_001287958.1
10
1065
NM_020886.3
NP_065937.1
12
1276
XM_005271630.2
XP_005271687.1
12
1275
XM_005271631.2
XP_005271688.1
12
1272
XM_005271632.2
XP_005271689.1
13
1605
XM_005271633.4
XP_005271690.1
13
1333
XM_005271636.2
XP_005271693.1
12
1275
XM_005271637.2
XP_005271694.1
12
1275
XM_005271638.2
XP_005271695.1
12
1275
XM_005271639.2
XP_005271696.1
12
1272
XM_011542936.2
XP_011541238.1
11
1194
XM_011542938.1
XP_011541240.1
10
1068
XM_011542941.1
XP_011541243.1
5
451
XM_017018056.1
XP_016873545.1
11
1191
XM_017018057.1
XP_016873546.1
13
1602
XM_017018058.1
XP_016873547.1
11
1208
XM_017018059.1
XP_016873548.1
11
1191
XM_017018060.1
XP_016873549.1
10
1065
XM_017018061.1
XP_016873550.1
10
1068
XM_017018062.1
XP_016873551.1
10
1065
XM_017018063.1
XP_016873552.1
11
1152
XM_017018064.1
XP_016873553.1
10
1073
XM_017018065.1
XP_016873554.1
13
1317
XM_017018066.1
XP_016873555.1
14
1679
XM_017018067.1
XP_016873556.1
9
928
XM_017018068.1
XP_016873557.1
5
451
GenBank mRNA
AB040948.1
-
3
419
AF266283.1
-
12
1213
AK097623.1
-
14
1389
AK304770.1
-
10
1036
BC065928.1
-
12
1234
6140725
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
DNA repair
ubiquitin-dependent protein catabolic process
cellular response to DNA damage stimulus
Ras protein signal transduction
cell proliferation
response to ionizing radiation
protein deubiquitination
regulation of protein stability
cellular response to UV
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
thiol-dependent ubiquitin-specific protease activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline