Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 5 | 714 | ||
NM_001815.4 | NP_001806.2 | 6 | 767 | |||
NR_102333.2 | 7 | 1271 | ||||
GenBank mRNA | BC106728.1 | - | 6 | 705 | 40015072 | |
D90277.1 | - | 6 | 753 | |||
D90278.1 | - | 7 | 1244 | |||
E03348.1 | - | 6 | 753 | |||
E03349.1 | - | 8 | 1126 | |||
L00692.1 | - | 6 | 733 | |||
L00693.1 | - | 5 | 680 |