Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199138.1 | NP_001186067.1 | 6 | 2687 | ||
NM_001199139.1 | NP_001186068.1 | 6 | 2570 | |||
NM_001302504.1 | NP_001289433.1 | 5 | 706 | |||
NM_021209.4 | NP_067032.3 | 6 | 2701 | |||
GenBank mRNA | AF376061.1 | - | 7 | 2915 | ||
AK095467.1 | - | 6 | 2687 | |||
AK292673.1 | - | 6 | 2672 | |||
AK314762.1 | - | 6 | 2672 | |||
AL389934.1 | - | 3 | 672 | |||
AY027787.1 | - | 6 | 2702 | |||
AY027788.1 | - | 5 | 707 | |||
AY032589.1 | - | 5 | 2461 | |||
AY035391.1 | - | 6 | 2570 | |||
AY358152.1 | - | 6 | 2649 | |||
BC031555.1 | - | 6 | 2657 | 5179909 |