Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021627.2 | NP_067640.2 | 11 | 1198 | ||
XM_005247690.2 | XP_005247747.2 | 11 | 1213 | |||
XM_005247691.2 | XP_005247748.1 | 11 | 1110 | |||
XM_017006973.1 | XP_016862462.1 | 11 | 1110 | |||
GenBank mRNA | AF151697.2 | - | 11 | 1201 | ||
AK027599.1 | - | 11 | 1139 | |||
AK074357.1 | - | 6 | 569 | |||
AK296744.1 | - | 8 | 743 | |||
AK296772.1 | - | 10 | 1079 | |||
AK298628.1 | - | 11 | 1025 | |||
AK304402.1 | - | 11 | 1267 | |||
AL834380.1 | - | 11 | 1022 | |||
BC040609.1 | - | 11 | 1084 | 5163937 |