Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001816.3 | NP_001807.2 | 4 | 1039 | ||
XM_011526340.2 | XP_011524642.1 | 4 | 1039 | |||
XM_011526341.1 | XP_011524643.1 | 4 | 861 | |||
XM_011526342.1 | XP_011524644.1 | 4 | 861 | |||
XM_017026194.1 | XP_016881683.1 | 4 | 1039 | |||
XM_017026195.1 | XP_016881684.1 | 4 | 1039 | |||
XM_017026196.1 | XP_016881685.1 | 4 | 1039 | |||
XM_017026197.1 | XP_016881686.1 | 3 | 765 | |||
XM_017026198.1 | XP_016881687.1 | 4 | 861 | |||
GenBank mRNA | AK297012.1 | - | 4 | 728 | ||
AK312652.1 | - | 4 | 1036 | |||
BC026263.1 | - | 4 | 1007 | 4618311 | ||
D90064.1 | - | 4 | 1023 | |||
M33326.1 | - | 4 | 1023 | |||
X52378.1 | - | 4 | 969 |