Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293557.1 | NP_001280486.1 | 11 | 4139 | ||
NM_022162.2 | NP_071445.1 | 12 | 4179 | |||
XM_005256084.3 | XP_005256141.1 | 12 | 4083 | |||
XM_006721242.3 | XP_006721305.1 | 11 | 3999 | |||
XM_011523259.2 | XP_011521561.1 | 10 | 3895 | |||
XM_017023535.1 | XP_016879024.1 | 10 | 5453 | |||
XM_017023536.1 | XP_016879025.1 | 11 | 3610 | |||
XM_017023537.1 | XP_016879026.1 | 10 | 3504 | |||
XM_017023538.1 | XP_016879027.1 | 11 | 3605 | |||
GenBank mRNA | AF178930.1 | - | 12 | 4179 |