Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022569.2 | NP_072091.1 | 2 | 1635 | ||
XM_017008546.1 | XP_016864035.1 | 1 | 27 | |||
GenBank mRNA | AB036429.1 | - | 2 | 1635 | ||
AK289670.1 | - | 2 | 1613 |