Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134223.1 | NP_001127695.1 | 9 | 2853 | ||
NM_001286767.1 | NP_001273696.1 | 9 | 2741 | |||
NM_001286768.1 | NP_001273697.1 | 10 | 2944 | |||
NM_022832.3 | NP_073743.2 | 9 | 2777 | |||
XM_017008557.1 | XP_016864046.1 | 9 | 2802 | |||
GenBank mRNA | AK024318.1 | - | 9 | 2718 | ||
BC037574.1 | - | 9 | 2726 | 5529667 |