Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042403.2 | NP_001035862.1 | 2 | 610 | ||
NM_001278393.1 | NP_001265322.1 | 2 | 826 | |||
NM_032147.4 | NP_115523.2 | 2 | 755 | |||
NR_125360.1 | 1 | 536 | ||||
XM_005269172.1 | XP_005269229.1 | 2 | 749 | |||
XM_005269173.1 | XP_005269230.1 | 2 | 826 | |||
XM_005269174.1 | XP_005269231.1 | 2 | 593 | |||
XM_011538800.2 | XP_011537102.1 | 3 | 776 | |||
XM_011538801.1 | XP_011537103.1 | 3 | 636 | |||
XM_011538802.1 | XP_011537104.1 | 2 | 596 | |||
XM_011538803.1 | XP_011537105.1 | 2 | 601 | |||
XM_011538804.2 | XP_011537106.1 | 2 | 809 | |||
XM_011538805.1 | XP_011537107.1 | 2 | 1450 | |||
XM_011538806.2 | XP_011537108.1 | 2 | 749 | |||
XM_017020009.1 | XP_016875498.1 | 2 | 744 | |||
XM_017020010.1 | XP_016875499.1 | 2 | 596 | |||
XM_017020011.1 | XP_016875500.1 | 3 | 622 | |||
XM_017020012.1 | XP_016875501.1 | 2 | 749 | |||
GenBank mRNA | AK027434.1 | - | 2 | 561 | ||
AK315697.1 | - | 2 | 758 | |||
AL136825.1 | - | 2 | 716 | |||
BC030704.1 | - | 2 | 591 | 4825887 |