Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290326.1 | NP_001277255.1 | 11 | 3755 | ||
NM_032557.6 | NP_115946.2 | 10 | 3639 | |||
XM_011532360.2 | XP_011530662.1 | 9 | 3025 | |||
GenBank mRNA | AB067478.1 | - | 9 | 2320 | ||
AF211481.1 | - | 2 | 1212 | |||
AK057992.1 | - | 7 | 2163 | |||
AK091712.1 | - | 3 | 1413 | |||
AK093289.1 | - | 2 | 689 | |||
AK093754.1 | - | 11 | 2978 | |||
AK126943.1 | - | 11 | 3467 | |||
AL833976.1 | - | 10 | 3611 | |||
BC068975.1 | - | 10 | 3586 | 6067501 |