Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032587.3 | NP_115976.2 | 1 | 290 | ||
XM_017009989.1 | XP_016865478.1 | 1 | 401 | |||
XM_017009990.1 | XP_016865479.1 | 1 | 408 | |||
GenBank mRNA | AB209844.1 | - | 1 | 110 | ||
AF356193.1 | - | 1 | 414 | |||
AK304349.1 | - | 1 | 222 | |||
AY196783.1 | - | 1 | 290 | |||
BC093825.1 | - | 1 | 237 | 7939670 |