Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHEX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000444.5 | NP_000435.3 | 2 | 707 | ||
NM_001282754.1 | NP_001269683.1 | 2 | 707 | |||
GenBank mRNA | AD000712.1 | - | 2 | 304 | ||
AK298030.1 | - | 2 | 253 | |||
AK301528.1 | - | 1 | 443 | |||
BC105057.1 | - | 2 | 229 | 8144060 | ||
BC105059.1 | - | 2 | 229 | 8144062 | ||
U60475.1 | - | 2 | 138 | |||
U75645.1 | - | 2 | 221 | |||
U82970.1 | - | 2 | 748 | |||
U87284.1 | - | 2 | 346 |