Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001868.3 | NP_001859.1 | 10 | 1246 | ||
GenBank mRNA | AK124421.1 | - | 3 | 1674 | ||
AK291493.1 | - | 10 | 1123 | |||
BC005279.1 | - | 10 | 1118 | 3949850 | ||
BT007313.1 | - | 10 | 1096 | |||
X67318.1 | - | 10 | 1103 |