Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003249.4 | NP_003240.1 | 2 | 346 | ||
GenBank mRNA | BC000135.2 | - | 2 | 272 | 2900551 | |
BC002391.2 | - | 2 | 298 | 2819803 | ||
BC013878.2 | - | 2 | 262 | 3690133 | ||
BU849088.1 | - | 2 | 346 | |||
U29366.1 | - | 2 | 230 | |||
Z50115.1 | - | 2 | 270 |