Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256063.1 | NP_001242992.1 | 4 | 337 | ||
NM_001256064.1 | NP_001242993.1 | 4 | 550 | |||
NM_001843.3 | NP_001834.2 | 4 | 337 | |||
NM_175038.2 | NP_778203.1 | 2 | 153 | |||
XM_005268651.2 | XP_005268708.1 | 4 | 700 | |||
XM_006719241.2 | XP_006719304.1 | 4 | 324 | |||
XM_011537926.2 | XP_011536228.1 | 4 | 289 | |||
XM_011537927.2 | XP_011536229.1 | 4 | 346 | |||
XM_017018821.1 | XP_016874310.1 | 4 | 340 | |||
XM_017018822.1 | XP_016874311.1 | 4 | 289 | |||
XM_017018823.1 | XP_016874312.1 | 4 | 700 | |||
XM_017018824.1 | XP_016874313.1 | 4 | 324 | |||
XM_017018825.1 | XP_016874314.1 | 4 | 289 | |||
XM_017018826.1 | XP_016874315.1 | 4 | 324 | |||
XM_017018827.1 | XP_016874316.1 | 4 | 346 | |||
GenBank mRNA | AK289544.1 | - | 4 | 429 | ||
AK289698.1 | - | 4 | 279 | |||
BC036569.2 | - | 4 | 276 | 5273941 | ||
U07819.1 | - | 3 | 110 | |||
U07820.1 | - | 2 | 158 | |||
Z21488.1 | - | 4 | 231 |