Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319116.1 | NP_001306045.1 | 3 | 321 | ||
NM_001319117.1 | NP_001306046.1 | 3 | 321 | |||
NM_001319118.1 | NP_001306047.1 | 3 | 249 | |||
NM_001319119.1 | NP_001306048.1 | 3 | 529 | |||
NM_001319120.1 | NP_001306049.1 | 3 | 346 | |||
NM_001319121.1 | NP_001306050.1 | 4 | 222 | |||
NM_001319122.1 | NP_001306051.1 | 3 | 314 | |||
NM_012100.3 | NP_036232.2 | 3 | 346 | |||
NR_134970.1 | 3 | 346 | ||||
GenBank mRNA | AF005050.2 | - | 3 | 321 | ||
AK001777.1 | - | 3 | 346 | |||
AK001977.1 | - | 3 | 318 | |||
AK021871.1 | - | 3 | 198 | |||
AK023005.1 | - | 3 | 260 | |||
AK301919.1 | - | 3 | 306 | |||
AK302809.1 | - | 3 | 313 | |||
BC000653.2 | - | 3 | 228 | 3343505 | ||
BI256589.1 | - | 3 | 212 | |||
BI821651.1 | - | 3 | 234 | |||
BM564135.1 | - | 3 | 309 | |||
BX403948.2 | - | 3 | 378 | |||
CN356162.1 | - | 3 | 240 | |||
DA203030.1 | - | 3 | 346 | |||
DA398660.1 | - | 3 | 249 |