Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163446.1 | NP_001156918.1 | 2 | 163 | ||
NM_016352.3 | NP_057436.2 | 2 | 163 | |||
GenBank mRNA | AF095719.1 | - | 2 | 123 | ||
AK290993.1 | - | 2 | 163 | |||
AK298550.1 | - | 2 | 134 | |||
AK298553.1 | - | 2 | 136 | |||
AK310933.1 | - | 2 | 136 | |||
AY358699.1 | - | 2 | 124 | |||
BC052289.1 | - | 2 | 144 | 6106874 | ||
DC368360.1 | - | 2 | 163 |