Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018226.4 | NP_060696.4 | 7, 8 | 1410 | ||
XM_005247036.4 | XP_005247093.1 | 7, 8 | 1471 | |||
GenBank mRNA | AF300795.1 | - | 7, 8 | 1116 | ||
AK001668.1 | - | 3, 4 | 374 | |||
AK130185.1 | - | 11 | ||||
BC017301.2 | - | 4, 5 | 562 | |||
BC067258.1 | - | 7, 8 | 985 | 6183046 | ||
BC082975.1 | - | 7, 8 | 1310 | 6379537 | ||
BC099906.1 | - | 5, 6 | 1523 | 5311883 |