Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035507.2 | NP_001030584.1 | 5 | 903 | ||
NM_021831.5 | NP_068603.4 | 5 | 903 | |||
NR_104246.1 | 5 | 903 | ||||
XM_005264477.2 | XP_005264534.1 | 5 | 903 | |||
XM_011533011.2 | XP_011531313.1 | 5 | 864 | |||
XM_011533012.2 | XP_011531314.1 | 6 | 1069 | |||
XM_011533013.2 | XP_011531315.1 | 6 | 872 | |||
GenBank mRNA | AK023398.1 | - | 5 | 858 | ||
AK025492.1 | - | 3 | 425 | |||
AK302091.1 | - | 5 | 834 | |||
AL833844.1 | - | 5 | 845 | |||
BC007415.2 | - | 5 | 845 | 2967512 | ||
BC018584.1 | - | 5 | 848 |