Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000171.3 | NP_000162.2 | 2 | 397 | ||
NM_001146040.1 | NP_001139512.1 | 2 | 397 | |||
XM_017009349.1 | XP_016864838.1 | 2 | 158 | |||
GenBank mRNA | AK308539.1 | - | 2 | 393 | ||
AK312702.1 | - | 2 | 323 | |||
BC074980.2 | - | 2 | 151 | 30915298 | ||
BC114947.1 | - | 2 | 397 | 40084102 | ||
DA749850.1 | - | 2 | 323 | |||
X52009.1 | - | 2 | 406 |