Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000824.4 | NP_000815.1 | 2 | 312 | ||
NM_001166060.1 | NP_001159532.1 | 2 | 295 | |||
NM_001166061.1 | NP_001159533.1 | 2 | 312 | |||
XM_017008035.1 | XP_016863524.1 | 2 | 289 | |||
GenBank mRNA | AF094754.1 | - | 2 | 275 | ||
AF094755.1 | - | 2 | 275 | |||
AK290617.1 | - | 2 | 246 | |||
BC032635.1 | - | 2 | 251 | 5247588 | ||
CD013911.1 | - | 1 | 149 | |||
DA168145.1 | - | 2 | 312 | |||
DA528659.1 | - | 2 | 312 | |||
DB479295.1 | - | 2 | 296 | |||
U33267.1 | - | 2 | 186 |