Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002240.4 | NP_002231.1 | 3 | 715 | ||
GenBank mRNA | AJ001894.1 | - | 3 | 416 | ||
AK313997.1 | - | 3 | 188 | |||
BC101547.1 | - | 3 | 188 | 8069053 | ||
D87327.1 | - | 3 | 636 | |||
U24660.1 | - | 3 | 345 |