Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278717.1 | NP_001265646.1 | 5 | 425 | ||
NM_001278718.1 | NP_001265647.1 | 5 | 319 | |||
NM_005031.4 | NP_005022.2 | 5 | 315 | |||
NM_021902.3 | NP_068702.1 | 5 | 291 | |||
XM_017026874.1 | XP_016882363.1 | 5 | 270 | |||
XM_017026875.1 | XP_016882364.1 | 5 | 485 | |||
XM_017026876.1 | XP_016882365.1 | 5 | 420 | |||
GenBank mRNA | AK289811.1 | - | 5 | 258 | ||
BC032800.2 | - | 4 | 185 | 5179163 | ||
BM691130.1 | - | 5 | 258 | |||
BM856555.1 | - | 5 | 252 | |||
BX115036.1 | - | 5 | 315 | |||
GQ258667.1 | - | 4 | 163 | |||
H57207.1 | - | 5 | 316 | |||
HY107209.1 | - | 5 | 292 | |||
HY111815.1 | - | 5 | 319 | |||
HY151101.1 | - | 5 | 280 | |||
HY151827.1 | - | 5 | 427 | |||
R72515.1 | - | 5 | 255 | |||
U72245.1 | - | 4 | 181 |