Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005549.2 | NP_005540.1 | 1 | 591 | ||
GenBank mRNA | BC074990.2 | - | 1 | 233 | 30915306 | |
CA309199.1 | - | 1 | 608 |