Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258448.1 | NP_001245377.1 | 4 | 414 | ||
NM_001258449.1 | NP_001245378.1 | 4 | 414 | |||
NM_001300921.1 | NP_001287850.1 | 4 | 414 | |||
NM_014386.3 | NP_055201.2 | 4 | 414 | |||
XM_011543318.2 | XP_011541620.1 | 3 | 328 | |||
XM_011543321.1 | XP_011541623.1 | 4 | 348 | |||
XM_017009343.1 | XP_016864832.1 | 4 | 430 | |||
XM_017009344.1 | XP_016864833.1 | 3 | 538 | |||
XM_017009345.1 | XP_016864834.1 | 4 | 430 | |||
GenBank mRNA | AF118125.1 | - | 4 | 381 | ||
AF182034.1 | - | 4 | 384 | |||
AK301924.1 | - | 4 | 389 | |||
BC034619.1 | - | 4 | 1467 | |||
BC038714.1 | - | 4 | 350 | |||
BC044581.1 | - | 4 | 413 | 4830805 | ||
BG719682.1 | - | 4 | 378 | |||
BG721989.1 | - | 4 | 419 |