Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001859.3 | NP_001850.1 | 2 | 300 | ||
GenBank mRNA | AK129613.1 | - | 2 | 242 | ||
AK222866.1 | - | 2 | 242 | |||
AK292511.1 | - | 2 | 254 | |||
AL831843.1 | - | 2 | 240 | |||
BC013611.1 | - | 2 | 229 | 3881191 | ||
BC061924.1 | - | 2 | 228 | 4802875 | ||
U83460.1 | - | 2 | 266 |