Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014505.5 | NP_055320.4 | 1, 2 | 797 | ||
XM_011538188.2 | XP_011536490.1 | 1, 2 | 859 | |||
GenBank mRNA | AF160967.1 | - | 1, 2 | 681 | ||
AF207992.1 | - | 1, 2 | 344 | |||
AF215891.1 | - | 1, 2 | 399 | |||
BC042446.2 | - | 1, 2 | 506 | 3873518 | ||
BC050621.1 | - | 1, 2 | 438 | 5552807 |