Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282534.1 | NP_001269463.1 | 2 | 1071 | ||
NR_104210.1 | 2 | 1071 | ||||
XM_011517102.2 | XP_011515404.1 | 2 | 1150 | |||
XM_017013530.1 | XP_016869019.1 | 2 | 866 | |||
XM_017013531.1 | XP_016869020.1 | 2 | 837 | |||
GenBank mRNA | AF212829.1 | - | 2 | 1007 | ||
AF248241.1 | - | 2 | 1071 | |||
AF257080.1 | - | 2 | 1013 | |||
AF279809.1 | - | 2 | 1007 | |||
AY190605.1 | - | 2 | 1007 | |||
BC075079.2 | - | 2 | 1056 | 30915596 | ||
BC075080.2 | - | 2 | 1056 | 30915383 | ||
BC112063.1 | - | 2 | 1056 | 8327531 | ||
BC112065.1 | - | 2 | 1056 | 8327533 |