Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182847.2 | NP_878267.2 | 1 | 1040 | ||
XM_005246671.2 | XP_005246728.2 | 1 | 897 | |||
GenBank mRNA | AK308804.1 | - | 1 | 897 | ||
AK309373.1 | - | 1 | 806 | |||
BC010439.1 | - | 1 | 880 | 4215164 | ||
BC031812.1 | - | 1 | 1040 | 4944238 |