Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000063.5 | NP_000054.2 | 6 | 1003 | ||
NM_001145903.2 | NP_001139375.1 | 4 | 607 | |||
NM_001282458.1 | NP_001269387.1 | 6 | 1031 | |||
NM_001282459.1 | NP_001269388.1 | 6 | 1003 | |||
GenBank mRNA | AK222537.1 | - | 6 | 755 | ||
AK222783.1 | - | 6 | 727 | |||
AK300892.1 | - | 4 | 367 | |||
AK300930.1 | - | 6 | 769 | |||
AK307930.1 | - | 4 | 363 | |||
AK312581.1 | - | 6 | 837 | |||
BC029781.1 | - | 6 | 777 | |||
BC043484.1 | - | 6 | 738 | 5763774 | ||
X04481.1 | - | 6 | 763 |