Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 3 | 357 | ||
NM_001319189.1 | NP_001306118.1 | 3 | 357 | |||
NM_033011.3 | NP_127509.1 | 3 | 357 | |||
GenBank mRNA | A03776.1 | - | 2 | 134 | ||
A07197.1 | - | 2 | 124 | |||
AK289387.1 | - | 3 | 309 | |||
AK297738.1 | - | 3 | 198 | |||
AK297937.1 | - | 3 | 198 | |||
AK300984.1 | - | 3 | 198 | |||
AK313342.1 | - | 3 | 198 | |||
AY221101.1 | - | 2 | 100 | |||
BC002795.2 | - | 3 | 183 | 3618149 | ||
BC007231.1 | - | 3 | 181 | 3161005 | ||
BC013968.2 | - | 3 | 183 | 3163742 | ||
BC018636.2 | - | 3 | 183 | 3637273 | ||
BC095403.1 | - | 3 | 198 | 30528441 | ||
BT007060.1 | - | 2 | 100 | |||
D01096.1 | - | 1 | 12 | |||
DA549782.1 | - | 3 | 309 | |||
FM956488.1 | - | 2 | 111 | |||
M15518.1 | - | 3 | 176 | |||
M18182.1 | - | 2 | 113 | |||
X02901.1 | - | 3 | 255 | |||
X07393.1 | - | 3 | 153 | |||
X13097.1 | - | 3 | 208 |