Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001909.4 | NP_001900.1 | 4 | 540 | ||
GenBank mRNA | AK130178.1 | - | 4 | 432 | ||
BC016320.2 | - | 4 | 452 | 3506977 | ||
BM762986.1 | - | 4 | 540 | |||
BT006910.1 | - | 4 | 407 | |||
BT020155.1 | - | 4 | 407 | |||
CR456947.1 | - | 4 | 407 | |||
EU794598.1 | - | 4 | 452 | |||
KU178007.1 | - | 4 | 407 | |||
M11233.1 | - | 4 | 458 | |||
X05344.1 | - | 4 | 409 |