Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CELA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001971.5 | NP_001962.3 | 5 | 458 | ||
GenBank mRNA | AF120493.1 | - | 5 | 458 | ||
BC069454.1 | - | 5 | 458 | 7262274 | ||
BC075091.2 | - | 5 | 439 | 30915390 |