Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166424.1 | NP_001159896.1 | 5 | 567 | ||
NM_002630.3 | NP_002621.1 | 5 | 567 | |||
GenBank mRNA | AK223274.1 | - | 5 | 563 | ||
AK301298.1 | - | 5 | 563 | |||
BC073740.1 | - | 5 | 549 | 6296399 | ||
J04443.1 | - | 5 | 507 | |||
U75272.1 | - | 5 | 550 |