Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002788.3 | NP_002779.1 | 4 | 374 | ||
NM_152132.2 | NP_687033.1 | 4 | 374 | |||
NR_038123.1 | 3 | 291 | ||||
GenBank mRNA | AK315158.1 | - | 4 | 323 | ||
BC005265.1 | - | 4 | 302 | 3996256 | ||
BC038990.1 | - | 4 | 301 | 6018899 | ||
BG703488.1 | - | 3 | 197 | |||
BT006711.1 | - | 4 | 258 | |||
BT019715.1 | - | 4 | 258 | |||
CF128376.1 | - | 4 | 374 | |||
CR456933.1 | - | 4 | 258 | |||
D00762.1 | - | 4 | 263 |