Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002801.3 | NP_002792.1 | 8 | 895 | ||
GenBank mRNA | AK225658.1 | - | 7 | 682 | ||
AK312208.1 | - | 8 | 855 | |||
BC017198.1 | - | 8 | 846 | 3143189 | ||
BC052369.1 | - | 8 | 833 | 4054165 | ||
BT019723.1 | - | 8 | 767 | |||
BT019724.1 | - | 8 | 767 | |||
CR456982.1 | - | 8 | 767 | |||
Y13640.1 | - | 8 | 855 |