Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004055.4 | NP_004046.2 | 6 | 1016 | ||
XM_011545225.1 | XP_011543527.1 | 7 | 1137 | |||
XM_017018223.1 | XP_016873712.1 | 6 | 939 | |||
GenBank mRNA | AB209087.1 | - | 6 | 945 | ||
AK094150.1 | - | 7 | 1088 | |||
AK310978.1 | - | 7 | 1057 | |||
AK313132.1 | - | 6 | 990 | |||
BC018123.1 | - | 6 | 1016 | 3904914 | ||
U94346.1 | - | 6 | 974 | |||
Y10552.1 | - | 5 | 874 |