Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199739.1 | NP_001186668.1 | 3 | 440 | ||
NM_004079.4 | NP_004070.3 | 3 | 440 | |||
GenBank mRNA | AK226095.1 | - | 3 | 302 | ||
AK301472.1 | - | 3 | 275 | |||
AK314482.1 | - | 3 | 281 | |||
BC002642.2 | - | 3 | 241 | 3610589 | ||
CR541676.1 | - | 2 | 179 | |||
M86553.1 | - | 2 | 185 | |||
M90696.1 | - | 3 | 315 | |||
S93414.1 | - | 2 | 185 |