Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004346.3 | NP_004337.2 | 4, 5 | 444 | ||
NM_032991.2 | NP_116786.1 | 3, 4 | 277 | |||
XM_011532301.1 | XP_011530603.1 | 4, 5 | 378 | |||
GenBank mRNA | AJ413269.1 | - | 2, 3 | 181 | ||
AK291337.1 | - | 4, 5 | 415 | |||
AU127028.1 | - | 3, 4 | 277 | |||
AU133051.1 | - | 4, 5 | 416 | |||
BC016926.2 | - | 4, 5 | 396 | 4419175 | ||
BX647609.1 | - | 4, 5 | 516 | |||
U13737.1 | - | 4, 5 | 405 | |||
U26943.1 | - | 2, 3 | 181 |