Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004651.3 | NP_004642.2 | 1 | 260 | ||
GenBank mRNA | AB073597.1 | - | 1 | 162 | ||
AK098394.1 | - | 1 | 164 | |||
BC000350.2 | - | 1 | 134 | |||
BC063668.1 | - | 1 | 142 | |||
BC140849.1 | - | 1 | 309 | 9021717 | ||
U44839.1 | - | 1 | 124 |