Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 6, 7 | 1094 | ||
NM_013421.2 | NP_038265.2 | 7, 8 | 1022 | |||
NM_013430.2 | NP_038347.2 | 6, 7 | 782 | |||
GenBank mRNA | AL832738.1 | - | 998 | |||
BC025927.1 | - | 4, 5 | 662 | 4548861 | ||
BU164348.1 | - | 6, 7 | 789 | |||
CR456494.1 | - | 3, 4 | 372 | |||
J04131.1 | - | 7, 8 | 964 | |||
J05235.1 | - | 5, 6 | 1039 | |||
M24087.1 | - | 2, 3 | 297 | |||
M24903.1 | - | 6, 7 | 782 | |||
X60069.1 | - | 4, 5 | 653 |