Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007272.2 | NP_009203.2 | 3 | 175 | ||
XM_011540550.1 | XP_011538852.1 | 3 | 175 | |||
GenBank mRNA | AK289447.1 | - | 3 | 174 | ||
BC015118.1 | - | 3 | 181 | 3950170 | ||
BC059779.1 | - | 3 | 154 | |||
S82198.1 | - | 3 | 152 |