Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 4 | 638 | ||
GenBank mRNA | AK290852.1 | - | 2 | 357 | ||
AK310424.1 | - | 2 | 311 | |||
BC018061.1 | - | 2 | 356 | 4817634 | ||
BI860684.1 | - | 3 | 311 | |||
BX648477.1 | - | 4 | 554 | |||
L02950.1 | - | 2 | 317 | |||
U85772.1 | - | 2 | 312 |