Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025104.1 | NP_001020275.1 | 5, 6 | 186 | ||
NM_001890.1 | NP_001881.1 | 5, 6 | 186 | |||
XM_006714089.2 | XP_006714152.1 | 5, 6 | 205 | |||
XM_006714090.2 | XP_006714153.1 | 5, 6 | 205 | |||
XM_006714091.2 | XP_006714154.1 | 5, 6 | 205 | |||
GenBank mRNA | BC128227.1 | - | 5, 6 | 175 | 40114618 | |
BC128228.1 | - | 5, 6 | 175 | 40114622 | ||
BG217918.1 | - | 4, 5 | 148 | |||
DQ064604.1 | - | 4, 5 | 137 | |||
U23157.1 | - | 5, 6 | 165 | |||
X78416.1 | - | 5, 6 | 186 | |||
X98084.1 | - | 4, 5 | 114 |