Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KREMEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039570.2 | NP_001034659.2 | 2 | 256 | ||
NM_032045.4 | NP_114434.3 | 2 | 256 | |||
XM_011530429.2 | XP_011528731.1 | 2 | 216 | |||
XM_011530430.2 | XP_011528732.1 | 2 | 218 | |||
XM_011530432.2 | XP_011528734.1 | 2 | 330 | |||
XM_017028989.1 | XP_016884478.1 | 2 | 218 | |||
GenBank mRNA | AB059618.1 | - | 2 | 197 | ||
AK056425.1 | - | 2 | 216 | |||
BC063787.1 | - | 2 | 268 | 6315614 |