Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178026.2 | NP_821158.2 | 3 | 559 | ||
XM_017027809.1 | XP_016883298.1 | 3 | 558 | |||
XM_017027810.1 | XP_016883299.1 | 3 | 558 | |||
XM_017027811.1 | XP_016883300.1 | 3 | 558 | |||
GenBank mRNA | AK097100.1 | - | 2 | 417 | ||
AK124788.1 | - | 2 | 873 | |||
AK299494.1 | - | 3 | 557 | |||
AY138815.1 | - | 3 | 516 | |||
BC033745.1 | - | 3 | 559 | |||
BC040030.1 | - | 4 | 574 | |||
BC101371.1 | - | 3 | 548 | |||
BC101372.1 | - | 3 | 555 | |||
BX401449.2 | - | 3 | 566 | |||
CA454346.1 | - | 3 | 555 |