Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031849.2 | NP_001027019.1 | 2 | 488 | ||
NM_001879.5 | NP_001870.3 | 2 | 488 | |||
NM_139125.3 | NP_624302.1 | 2 | 488 | |||
XM_006713701.2 | XP_006713764.1 | 3 | 561 | |||
XM_011512989.2 | XP_011511291.1 | 2 | 482 | |||
XM_011512990.2 | XP_011511292.1 | 2 | 482 | |||
XM_011512991.2 | XP_011511293.1 | 2 | 477 | |||
XM_017006869.1 | XP_016862358.1 | 3 | 566 | |||
XM_017006871.1 | XP_016862360.1 | 2 | 482 | |||
XM_017006872.1 | XP_016862361.1 | 1 | 71 | |||
GenBank mRNA | AF284421.1 | - | 2 | 188 | ||
AK129893.1 | - | 2 | 187 | |||
AK291157.1 | - | 2 | 324 | |||
AK291686.1 | - | 2 | 166 | |||
AK300663.1 | - | 3 | 309 | |||
AL134380.1 | - | 2 | 430 | |||
BC039724.1 | - | 2 | 423 | |||
BC106945.1 | - | 2 | 148 | 40034551 | ||
BC106946.1 | - | 2 | 148 | 40034552 | ||
CR749615.1 | - | 2 | 390 | |||
DC423174.1 | - | 2 | 488 |