Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152891.2 | NP_690851.2 | 6 | 1377 | ||
XM_011522451.2 | XP_011520753.1 | 7 | 1501 | |||
XM_011522452.2 | XP_011520754.1 | 7 | 1313 | |||
GenBank mRNA | AF536382.1 | - | 6 | 1284 | ||
BC036846.1 | - | 7 | 1359 | 5245847 | ||
BC062334.1 | - | 6 | 1379 | 6110198 |