Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153692.3 | NP_710159.1 | 4 | 923 | ||
XM_011544431.2 | XP_011542733.1 | 4 | 923 | |||
XM_011544432.1 | XP_011542734.1 | 4 | 923 | |||
GenBank mRNA | AK075205.1 | - | 4 | 894 | ||
AK300197.1 | - | 4 | 802 | |||
BC057765.1 | - | 4 | 855 | 30348811 |